Assay Details
| Gene Symbol: dpysl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005637.1 | NP_001005637.1 | 7 - 8 | 752 | 66 | |
| XM_012958802.2 | XP_012814256.1 | 7 - 8 | 1028 | 66 | ||
| XM_012958803.2 | XP_012814257.1 | 6 - 7 | 947 | 66 | ||
| GenBank mRNA | BC074633.1 | - | 7 - 8 | 752 | 66 | |
| CR942629.1 | - | 7 - 8 | 753 | 66 | ||
More Information
Set Membership: |
|

dpysl3
Genomic Map