Assay Details
| Gene Symbol: krt5.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005638.1 | NP_001005638.1 | 5 - 6 | 484 | 96 | |
| XM_012968195.2 | XP_012823649.1 | 5 - 6 | 561 | 96 | ||
| XM_012968196.2 | XP_012823650.1 | 5 - 6 | 532 | 96 | ||
| XM_012968197.2 | XP_012823651.1 | 5 - 6 | 563 | 96 | ||
| GenBank mRNA | BC074634.1 | - | 5 - 6 | 484 | 96 | |
| CR855564.2 | - | 5 - 6 | 476 | 96 | ||
| CR926161.1 | - | 4 - 5 | 310 | 96 | ||
More Information
Set Membership: |
|

krt5.1
Genomic Map