Assay Details
| Gene Symbol: krt5.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005638.1 | NP_001005638.1 | 7 - 8 | 670 | 87 | |
| XM_012968195.2 | XP_012823649.1 | 7 - 8 | 747 | 87 | ||
| XM_012968196.2 | XP_012823650.1 | 7 - 8 | 718 | 87 | ||
| XM_012968197.2 | XP_012823651.1 | 7 - 8 | 749 | 87 | ||
| GenBank mRNA | BC074634.1 | - | 7 - 8 | 670 | 87 | |
| CR855564.2 | - | 7 - 8 | 662 | 87 | ||
| CR926161.1 | - | 6 - 7 | 496 | 87 | ||
| Gene Symbol: LOC100489158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002935728.4 | XP_002935774.1 | 3342 | 87 | ||
More Information
Set Membership: |
|

krt5.1
Genomic Map