Assay Details
| Gene Symbol: rassf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005644.1 | NP_001005644.1 | 4 - 5 | 906 | 68 | |
| XM_012958805.2 | XP_012814259.1 | 5 - 6 | 1002 | 68 | ||
| XM_012958806.1 | XP_012814260.1 | 5 - 6 | 901 | 68 | ||
| GenBank mRNA | BC074644.1 | - | 4 - 5 | 906 | 68 | |
| CR942435.3 | - | 4 - 5 | 651 | 68 | ||
More Information
Set Membership: |
|

rassf3
Genomic Map