Assay Details
| Gene Symbol: hnrnph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005664.1 | NP_001005664.1 | 6 - 7 | 768 | 100 | |
| XM_018092018.1 | XP_017947507.1 | 4 - 5 | 562 | 100 | ||
| GenBank mRNA | BC074690.1 | - | 6 - 7 | 768 | 100 | |
| CR760556.2 | - | 5 - 6 | 749 | 100 | ||
More Information
Set Membership: |
|

hnrnph1
Genomic Map