Assay Details
| Gene Symbol: pgam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005665.1 | NP_001005665.1 | 2 - 3 | 682 | 78 | |
| GenBank mRNA | BC074692.1 | - | 2 - 3 | 682 | 78 | |
More Information
Set Membership: |
|

pgam2
Genomic Map