Assay Details
| Gene Symbol: pip4k2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005670.1 | NP_001005670.1 | 5 - 6 | 765 | 69 | |
| XM_018091158.1 | XP_017946647.1 | 5 - 6 | 882 | 69 | ||
| GenBank mRNA | BC074705.1 | - | 5 - 6 | 765 | 69 | |
| CR762295.2 | - | 5 - 6 | 705 | 69 | ||
More Information
Set Membership: |
|

pip4k2c
Genomic Map