Assay Details
| Gene Symbol: g6pc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005682.1 | NP_001005682.1 | 1 - 2 | 267 | 75 | |
| GenBank mRNA | BC075103.1 | - | 1 - 2 | 267 | 75 | |
More Information
Set Membership: |
|

g6pc
Genomic Map