Assay Details
| Gene Symbol: epc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005683.1 | NP_001005683.1 | 12 - 13 | 2240 | 72 | |
| GenBank mRNA | BC075105.1 | - | 12 - 13 | 2240 | 72 | |
More Information
Set Membership: |
|

epc2
Genomic Map