Assay Details
| Gene Symbol: hccs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005708.2 | NP_001005708.2 | 4 - 5 | 609 | 118 | |
| NM_001247977.1 | NP_001234906.1 | 4 - 5 | 658 | 118 | ||
| NM_001247978.1 | NP_001234907.1 | 4 - 5 | 612 | 118 | ||
| XM_012960825.2 | XP_012816279.1 | 4 - 5 | 599 | 118 | ||
| GenBank mRNA | BC075286.1 | - | 4 - 5 | 542 | 118 | |
| CR926219.2 | - | 3 - 4 | 520 | 118 | ||
| CX505765.2 | - | 4 - 5 | 641 | 118 | ||
| CX934649.1 | - | 4 - 5 | 601 | 118 | ||
| DT398620.1 | - | 4 - 5 | 607 | 118 | ||
| EL797799.1 | - | 3 - 4 | 603 | 118 | ||
More Information
Set Membership: |
|

hccs
Genomic Map