Assay Details
| Gene Symbol: naaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005714.1 | NP_001005714.1 | 6 - 7 | 910 | 80 | |
| GenBank mRNA | BC075301.1 | - | 6 - 7 | 910 | 80 | |
More Information
Set Membership: |
|

naaa
Genomic Map