Assay Details
| Gene Symbol: lcat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005715.1 | NP_001005715.1 | 1 - 2 | 230 | 81 | |
| XM_018092991.1 | XP_017948480.1 | 2 - 3 | 286 | 81 | ||
| GenBank mRNA | BC075304.1 | - | 1 - 2 | 230 | 81 | |
More Information
Set Membership: |
|

lcat
Genomic Map