Assay Details
| Gene Symbol: lcat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005715.1 | NP_001005715.1 | 4 - 5 | 600 | 83 | |
| XM_018092991.1 | XP_017948480.1 | 5 - 6 | 656 | 83 | ||
| GenBank mRNA | BC075304.1 | - | 4 - 5 | 600 | 83 | |
More Information
Set Membership: |
|

lcat
Genomic Map