Assay Details
| Gene Symbol: babam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005716.1 | NP_001005716.1 | 5 - 6 | 543 | 79 | |
| XM_018093838.1 | XP_017949327.1 | 6 - 7 | 783 | 79 | ||
| GenBank mRNA | BC075305.1 | - | 5 - 6 | 543 | 79 | |
More Information
Set Membership: |
|

babam2
Genomic Map