Assay Details
| Gene Symbol: mest | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005789.1 | NP_001005789.1 | 9 - 10 | 776 | 65 | |
| XM_012958824.2 | XP_012814278.1 | 9 - 10 | 828 | 65 | ||
| XM_018092023.1 | XP_017947512.1 | 10 - 11 | 1055 | 65 | ||
| GenBank mRNA | BC075327.1 | - | 9 - 10 | 776 | 65 | |
More Information
Set Membership: |
|

mest
Genomic Map