Assay Details
| Gene Symbol: pdlim3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005814.1 | NP_001005814.1 | 2 - 3 | 323 | 61 | |
| XM_012959429.2 | XP_012814883.1 | 2 - 3 | 436 | 61 | ||
| XM_012959432.2 | XP_012814886.1 | 2 - 3 | 434 | 61 | ||
| XM_012959434.2 | XP_012814888.1 | 2 - 3 | 428 | 61 | ||
| GenBank mRNA | BC075363.1 | - | 2 - 3 | 323 | 61 | |
More Information
Set Membership: |
|

pdlim3
Genomic Map