Assay Details
| Gene Symbol: htatip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005817.1 | NP_001005817.1 | 3 - 4 | 406 | 90 | |
| XM_012960840.2 | XP_012816294.1 | 3 - 4 | 448 | 90 | ||
| XM_012960841.2 | XP_012816295.1 | 3 - 4 | 394 | 90 | ||
| GenBank mRNA | BC075366.1 | - | 3 - 4 | 406 | 90 | |
More Information
Set Membership: |
|

htatip2
Genomic Map