Assay Details
| Gene Symbol: bglap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006688.1 | NP_001006689.1 | 3 - 4 | 202 | 97 | |
| GenBank mRNA | BC075385.1 | - | 3 - 4 | 202 | 97 | |
More Information
Set Membership: |
|

bglap
Genomic Map