Assay Details
| Gene Symbol: ankra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006691.1 | NP_001006692.1 | 1 - 2 | 347 | 60 | |
| XM_012959488.2 | XP_012814942.1 | 2 - 3 | 455 | 60 | ||
| XM_012959498.2 | XP_012814952.1 | 2 - 3 | 1061 | 60 | ||
| XM_012959503.2 | XP_012814957.1 | 2 - 3 | 492 | 60 | ||
| XM_012959507.2 | XP_012814961.1 | 3 - 4 | 1011 | 60 | ||
| XM_012959516.2 | XP_012814970.1 | 2 - 3 | 390 | 60 | ||
| GenBank mRNA | BC075392.1 | - | 1 - 2 | 347 | 60 | |
More Information
Set Membership: |
|

ankra2
Genomic Map