Assay Details
| Gene Symbol: nek6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006699.1 | NP_001006700.1 | 10 - 11 | 1164 | 68 | |
| XM_012953020.2 | XP_012808474.1 | 11 - 12 | 1366 | 68 | ||
| XM_012953021.2 | XP_012808475.1 | 9 - 10 | 995 | 68 | ||
| GenBank mRNA | BC075406.1 | - | 10 - 11 | 1164 | 68 | |
| CR760768.2 | - | 10 - 11 | 1166 | 68 | ||
More Information
Set Membership: |
|

nek6
Genomic Map