Assay Details
| Gene Symbol: agxt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006704.1 | NP_001006705.1 | 10 - 11 | 1257 | 57 | |
| GenBank mRNA | BC075417.1 | - | 10 - 11 | 1257 | 57 | |
| BC089751.1 | - | 10 - 11 | 1353 | 57 | ||
More Information
Set Membership: |
|

agxt
Genomic Map