Assay Details
| Gene Symbol: arg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006713.1 | NP_001006714.1 | 3 - 4 | 320 | 68 | |
| GenBank mRNA | BC075440.1 | - | 3 - 4 | 320 | 68 | |
More Information
Set Membership: |
|

arg1
Genomic Map