Assay Details
| Gene Symbol: myoz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006716.1 | NP_001006717.1 | 3 - 4 | 429 | 69 | |
| XM_012966388.2 | XP_012821842.1 | 3 - 4 | 451 | 69 | ||
| GenBank mRNA | BC075445.1 | - | 3 - 4 | 429 | 69 | |
More Information
Set Membership: |
|

myoz1
Genomic Map