Assay Details
| Gene Symbol: znf534 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006744.1 | NP_001006745.1 | 2 - 3 | 1115 | 92 | |
| XM_012968223.2 | XP_012823677.1 | 2 - 3 | 1173 | 92 | ||
| GenBank mRNA | BC075511.1 | - | 2 - 3 | 1115 | 92 | |
More Information
Set Membership: |
|

znf534
Genomic Map