Assay Details
| Gene Symbol: stk38l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006752.3 | NP_001006753.1 | 193 | 59 | ||
| GenBank mRNA | BC075525.1 | - | 2 - 3 | 235 | 59 | |
More Information
Set Membership: |
|

stk38l
Genomic Map