Assay Details
| Gene Symbol: spa17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006757.1 | NP_001006758.1 | 7 - 8 | 1591 | 72 | |
| XM_012966399.2 | XP_012821853.1 | 7 - 8 | 1862 | 72 | ||
| XM_012966400.2 | XP_012821854.1 | 7 - 8 | 1977 | 72 | ||
| XM_012966402.2 | XP_012821856.1 | 6 - 7 | 1740 | 72 | ||
| GenBank mRNA | BC075539.1 | - | 7 - 8 | 1591 | 72 | |
| CT030312.1 | - | 1641 | 72 | |||
More Information
Set Membership: |
|

spa17
Genomic Map