Assay Details
| Gene Symbol: usp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006760.1 | NP_001006761.1 | 10 - 11 | 1729 | 71 | |
| XM_012960855.2 | XP_012816309.2 | 9 - 10 | 1788 | 71 | ||
| XM_018092993.1 | XP_017948482.1 | 11 - 12 | 1915 | 71 | ||
| GenBank mRNA | BC075544.1 | - | 10 - 11 | 1729 | 71 | |
More Information
Set Membership: |
|

usp10
Genomic Map