Assay Details
| Gene Symbol: pax3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006775.2 | NP_001006776.2 | 7 - 8 | 1281 | 60 | |
| XM_012970301.2 | XP_012825755.1 | 7 - 8 | 1852 | 60 | ||
| XM_012970302.2 | XP_012825756.1 | 7 - 8 | 1849 | 60 | ||
| XM_012970303.2 | XP_012825757.1 | 7 - 8 | 1837 | 60 | ||
| XM_012970304.2 | XP_012825758.1 | 7 - 8 | 1852 | 60 | ||
| XM_012970306.2 | XP_012825760.1 | 7 - 8 | 1777 | 60 | ||
| XM_012970307.2 | XP_012825761.1 | 7 - 8 | 1762 | 60 | ||
| GenBank mRNA | BC075577.1 | - | 7 - 8 | 1345 | 60 | |
| CR855399.2 | - | 7 - 8 | 1281 | 60 | ||
More Information
Set Membership: |
|

pax3
Genomic Map