Assay Details
| Gene Symbol: taf5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006778.1 | NP_001006779.1 | 4 - 5 | 1143 | 129 | |
| XM_018093841.1 | XP_017949330.1 | 3 - 4 | 1116 | 129 | ||
| GenBank mRNA | BC075582.1 | - | 4 - 5 | 1143 | 129 | |
| CR761718.2 | - | 4 - 5 | 1208 | 129 | ||
More Information
Set Membership: |
|

taf5l
Genomic Map