Assay Details
| Gene Symbol: syngr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006784.1 | NP_001006785.1 | 3 - 4 | 577 | 81 | |
| GenBank mRNA | BC075595.1 | - | 3 - 4 | 577 | 81 | |
More Information
Set Membership: |
|

syngr2
Genomic Map