Assay Details
| Gene Symbol: gch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006788.1 | NP_001006789.1 | 5 - 6 | 714 | 62 | |
| GenBank mRNA | BC075602.1 | - | 5 - 6 | 714 | 62 | |
More Information
Set Membership: |
|

gch1
Genomic Map