Assay Details
| Gene Symbol: entpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006795.1 | NP_001006796.1 | 3 - 4 | 547 | 63 | |
| XM_018095347.1 | XP_017950836.1 | 3 - 4 | 608 | 63 | ||
| XM_018095348.1 | XP_017950837.1 | 2 - 3 | 653 | 63 | ||
| GenBank mRNA | BC076662.1 | - | 3 - 4 | 547 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018095348.1 transcripts.
Set Membership: |
|

entpd1
Genomic Map