Assay Details
| Gene Symbol: ufd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006805.1 | NP_001006806.1 | 5 - 6 | 445 | 85 | |
| XM_018092252.1 | XP_017947741.1 | 5 - 6 | 445 | 85 | ||
| GenBank mRNA | BC076680.1 | - | 5 - 6 | 445 | 85 | |
More Information
Set Membership: |
|

ufd1
Genomic Map