Assay Details
| Gene Symbol: gpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006807.1 | NP_001006808.1 | 2 - 3 | 267 | 62 | |
| GenBank mRNA | BC076683.1 | - | 2 - 3 | 267 | 62 | |
| CR761150.2 | - | 2 - 3 | 289 | 62 | ||
More Information
Set Membership: |
|

gpd1
Genomic Map