Assay Details
| Gene Symbol: prdx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006811.1 | NP_001006812.1 | 6 - 7 | 836 | 56 | |
| GenBank mRNA | BC076692.1 | - | 6 - 7 | 836 | 56 | |
| CR762126.2 | - | 6 - 7 | 790 | 56 | ||
More Information
Set Membership: |
|

prdx4
Genomic Map