Assay Details
| Gene Symbol: arpc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006823.1 | NP_001006824.1 | 4 - 5 | 528 | 74 | |
| GenBank mRNA | BC076887.1 | - | 4 - 5 | 528 | 74 | |
| CR760485.2 | - | 4 - 5 | 451 | 74 | ||
More Information
Set Membership: |
|

arpc1b
Genomic Map