Assay Details
| Gene Symbol: clic4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006830.1 | NP_001006831.1 | 2 - 3 | 429 | 75 | |
| XM_012970309.1 | XP_012825763.1 | 3 - 4 | 683 | 75 | ||
| GenBank mRNA | BC076899.1 | - | 2 - 3 | 429 | 75 | |
More Information
Set Membership: |
|

clic4
Genomic Map