Assay Details
| Gene Symbol: hal.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006831.2 | NP_001006832.2 | 14 - 15 | 1382 | 118 | |
| NM_001278528.1 | NP_001265457.1 | 15 - 16 | 1401 | 118 | ||
| XM_012958847.2 | XP_012814301.1 | 15 - 16 | 1537 | 118 | ||
| XM_012958848.1 | XP_012814302.1 | 15 - 16 | 1350 | 118 | ||
| XM_012958849.2 | XP_012814303.1 | 15 - 16 | 1417 | 118 | ||
| XM_012958850.1 | XP_012814304.1 | 14 - 15 | 1229 | 118 | ||
| XM_012958851.1 | XP_012814305.1 | 14 - 15 | 1108 | 118 | ||
| XM_018092036.1 | XP_017947525.1 | 15 - 16 | 1350 | 118 | ||
| XM_018092037.1 | XP_017947526.1 | 14 - 15 | 1312 | 118 | ||
| GenBank mRNA | BC076901.1 | - | 14 - 15 | 1381 | 118 | |
| DN034118.1 | - | 2 - 3 | 104 | 118 | ||
| DN035766.1 | - | 4 - 5 | 263 | 118 | ||
More Information
Set Membership: |
|

hal.2
Genomic Map