Assay Details
| Gene Symbol: hal.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006831.2 | NP_001006832.2 | 1 - 2 | 347 | 107 | |
| NM_001278528.1 | NP_001265457.1 | 2 - 3 | 366 | 107 | ||
| XM_012958847.2 | XP_012814301.1 | 2 - 3 | 502 | 107 | ||
| XM_012958848.1 | XP_012814302.1 | 2 - 3 | 315 | 107 | ||
| XM_012958849.2 | XP_012814303.1 | 2 - 3 | 382 | 107 | ||
| XM_018092036.1 | XP_017947525.1 | 2 - 3 | 315 | 107 | ||
| XM_018092037.1 | XP_017947526.1 | 2 - 3 | 403 | 107 | ||
| GenBank mRNA | BC076901.1 | - | 1 - 2 | 346 | 107 | |
| CF343491.1 | - | 1 - 2 | 363 | 107 | ||
| DN033227.1 | - | 2 - 3 | 366 | 107 | ||
More Information
Set Membership: |
|

hal.2
Genomic Map