Assay Details
| Gene Symbol: hal.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006831.2 | NP_001006832.2 | 9 - 10 | 958 | 95 | |
| NM_001278528.1 | NP_001265457.1 | 10 - 11 | 977 | 95 | ||
| XM_012958847.2 | XP_012814301.1 | 10 - 11 | 1113 | 95 | ||
| XM_012958848.1 | XP_012814302.1 | 10 - 11 | 926 | 95 | ||
| XM_012958849.2 | XP_012814303.1 | 10 - 11 | 993 | 95 | ||
| XM_012958850.1 | XP_012814304.1 | 9 - 10 | 805 | 95 | ||
| XM_012958851.1 | XP_012814305.1 | 9 - 10 | 684 | 95 | ||
| XM_018092036.1 | XP_017947525.1 | 10 - 11 | 926 | 95 | ||
| XM_018092037.1 | XP_017947526.1 | 9 - 10 | 888 | 95 | ||
| GenBank mRNA | BC076901.1 | - | 9 - 10 | 957 | 95 | |
| DN027070.1 | - | 6 - 7 | 482 | 95 | ||
More Information
Set Membership: |
|

hal.2
Genomic Map