Assay Details
| Gene Symbol: gnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006834.1 | NP_001006835.1 | 9 - 10 | 1388 | 84 | |
| XM_012966430.2 | XP_012821884.1 | 9 - 10 | 1047 | 84 | ||
| XM_012966431.2 | XP_012821885.1 | 9 - 10 | 1147 | 84 | ||
| GenBank mRNA | BC076910.1 | - | 9 - 10 | 1388 | 84 | |
More Information
Set Membership: |
|

gnb1
Genomic Map