Assay Details
| Gene Symbol: csrp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006835.1 | NP_001006836.1 | 2 - 3 | 171 | 63 | |
| XM_012960867.2 | XP_012816321.1 | 2 - 3 | 224 | 63 | ||
| GenBank mRNA | BC076911.1 | - | 2 - 3 | 171 | 63 | |
More Information
Set Membership: |
|

csrp3
Genomic Map