Assay Details
| Gene Symbol: txnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006843.1 | NP_001006844.1 | 1 - 2 | 179 | 62 | |
| GenBank mRNA | BC076929.1 | - | 1 - 2 | 179 | 62 | |
More Information
Set Membership: |
|

txnl1
Genomic Map