Assay Details
| Gene Symbol: pemt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006844.1 | NP_001006845.1 | 6 - 7 | 730 | 67 | |
| GenBank mRNA | BC076930.1 | - | 6 - 7 | 730 | 67 | |
| CT485695.1 | - | 6 - 7 | 770 | 67 | ||
More Information
Set Membership: |
|

pemt
Genomic Map