Assay Details
| Gene Symbol: uxs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006848.1 | NP_001006849.1 | 8 - 9 | 750 | 69 | |
| GenBank mRNA | BC076935.1 | - | 8 - 9 | 750 | 69 | |
| CT027882.1 | - | 8 - 9 | 650 | 69 | ||
More Information
Set Membership: |
|

uxs1
Genomic Map