Assay Details
| Gene Symbol: kif2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006855.1 | NP_001006856.1 | 20 - 21 | 2292 | 64 | |
| GenBank mRNA | BC076961.2 | - | 20 - 21 | 2292 | 64 | |
| CR855595.2 | - | 20 - 21 | 2155 | 64 | ||
More Information
Set Membership: |
|

kif2c
Genomic Map