Assay Details
| Gene Symbol: mycn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006873.1 | NP_001006874.1 | 2 - 3 | 1038 | 119 | |
| GenBank mRNA | BC076996.1 | - | 2 - 3 | 1038 | 119 | |
| CR761260.2 | - | 2 - 3 | 969 | 116 | ||
More Information
Set Membership: |
|

mycn
Genomic Map