Assay Details
| Gene Symbol: csrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006880.2 | NP_001006881.1 | 5 - 6 | 620 | 72 | |
| XM_012960188.2 | XP_012815642.1 | 6 - 7 | 705 | 72 | ||
| XM_018091175.1 | XP_017946664.1 | 6 - 7 | 707 | 72 | ||
| GenBank mRNA | BC077045.1 | - | 5 - 6 | 621 | 72 | |
More Information
Set Membership: |
|

csrp1
Genomic Map