Assay Details
| Gene Symbol: pxmp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006884.2 | NP_001006885.1 | 4 - 5 | 554 | 74 | |
| XM_018092416.1 | XP_017947905.1 | 4 - 5 | 451 | 74 | ||
| GenBank mRNA | BC077678.1 | - | 4 - 5 | 555 | 74 | |
| CT030300.1 | - | 4 - 5 | 547 | 74 | ||
More Information
Set Membership: |
|

pxmp2
Genomic Map