Assay Details
| Gene Symbol: srp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006888.1 | NP_001006889.1 | 2 - 3 | 137 | 84 | |
| GenBank mRNA | BC077688.1 | - | 2 - 3 | 137 | 84 | |
| CR855756.2 | - | 2 - 3 | 115 | 84 | ||
More Information
Set Membership: |
|

srp14
Genomic Map